Learn More
Cole-Parmer Idex Flangeless Fitting, Standard Knurl, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK
Catalog No. NC2101628 Shop All Cole-Parmer Products
Encompass
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components.
This item is not returnable. View return policy
Catalog No. NC2101628 Supplier Cole-Parmer Supplier No. 0201503
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components.