Learn More
AVANTOR MASTERFLEX (VWR LLC) FTNG LB FLRXB 3/16 CVU 10PK
Catalog No. 502742497
Encompass
Masterflex Large Bore Female Luer 3/16IN ID in Polypropylene Cleanroom Manufactured, Avantor
Catalog No. 50-274-2497 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4031005
Masterflex Large Bore Female Luer 3/16IN ID in Polypropylene Cleanroom Manufactured, Avantor