Learn More
AVANTOR MASTERFLEX (VWR LLC) FTNG MLR SLPXB 1/16 PPQC 10PK
Catalog No. 502742432
Encompass
Masterflex Male Slip Luer 1/16IN ID in Polypropylene Cleanroom Manufactured, Avantor
Catalog No. 50-274-2432 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4032109
Masterflex Male Slip Luer 1/16IN ID in Polypropylene Cleanroom Manufactured, Avantor