Learn More
AVANTOR MASTERFLEX (VWR LLC) FTNG MLRXB 3/16 PVDFQC 10PK
Catalog No. 502742449
Encompass
Masterflex Male Luer 3/16IN ID Barb in Natural PVDF Cleanroom Manufactured, Avantor
Catalog No. 50-274-2449 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4031822
Masterflex Male Luer 3/16IN ID Barb in Natural PVDF Cleanroom Manufactured, Avantor