Learn More
AVANTOR MASTERFLEX (VWR LLC) FTNG R MLRXB 1/8 PP 10PK
Catalog No. 502742439
Encompass
Masterflex Male Rotating Luer Lock and Nut Assembly 1/8IN ID in Polypropylene
Catalog No. 50-274-2439 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4032011
Masterflex Male Rotating Luer Lock and Nut Assembly 1/8IN ID in Polypropylene