Learn More
Aviva Systems Biology Corp FTO Antibody - middle region (ARP47474_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47474_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FTO. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human FTO. Alternate names: KIAA1752, MGC5149ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-309