Learn More
Aviva Systems Biology Corp Fto Antibody - N-terminal region (ARP47473_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47473_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Fto. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AW743446, mKIAA1752ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Yeast
Catalog No. 50-285-308