Learn More
Bioss FTO/Fat mass and obesity relat
SDP
Catalog No. 5000065137
Encompass_Preferred
FTO (Fat mass and obesity related protein) Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Clonality Monoclonal Applications WB 5-2H10Concentration Lot Dependent Size 100 ul Storage Buffer 10 mM HEPES (pH 7 5) 150 mM NaCl 100 g per ml BSA and 50% glycerol
Catalog No. 50-000-65137 Supplier Bioss Supplier No. BSM70094M
FTO (Fat mass and obesity related protein) Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Clonality Monoclonal Applications WB 5-2H10Concentration Lot Dependent Size 100 ul Storage Buffer 10 mM HEPES (pH 7 5) 150 mM NaCl 100 g per ml BSA and 50% glycerol