Learn More
AdipoGen anti-FTO (human), mAb (AG103) (50 µg)
SDP
Supplier:  AdipoGen AG20A0092C050
Encompass_Preferred
Monoclonal Antibody. Recognizes human FTO. Isotype: Mouse IgG2akappa. Clone: AG103. Applications: ELISA, IHC, IP, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. FTO (Fat mass-and obesity-associated gene) is the responsible gene for mouse ‘fused toes’ mutation. An association between FTO genotype and type 2 diabetes has been confirmed. The presence of the FTO rs9939609 A-allele was found to be positively correlated with other symptoms of the metabolic syndrome, including higher fasting insulin, glucose, triglycerides, and lower HDL-cholesterol.
Catalog No. 50-205-2894