Learn More
AdipoGen FTO human pAb
SDP
Supplier:  AdipoGen AG25A0084C100
Encompass_Preferred
Polyclonal Antibody. Recognizes human FTO. Detects a band of ~59kDa by Western blot. Source Rabbit. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. FTO Fat mass-and obesity-associated gene is the responsible gene for mouse ‘fused toes’ mutation. An association between FTO genotype and type 2 diabetes has been confirmed. The presence of the FTO rs9939609 A-allele was found to be positively correlated with other symptoms of the metabolic syndrome, including higher fasting insulin, glucose, triglycerides, and lower HDL-cholesterol.
Catalog No. 50-256-3233