Learn More
AdipoGen FTO human rec His
SDP
Catalog No. 502562993
Encompass_Preferred
Protein. Human FTO aa 2-505 is fused at the N-terminus to a His-tag. Source E. coli. Endotoxin content <1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in 55mM TRIS-Cl, pH 8.2, containing 150mM NaCl. Purity >90% SDS-PAGE. FTO Fat mass-and obesity-associated gene is the responsible gene for mouse ‘fused toes’ mutation. An association between FTO genotype and type 2 diabetes has been confirmed. The presence of the FTO rs9939609 A-allele was found to be positively correlated with other symptoms of the metabolic syndrome, including higher fasting insulin, glucose, triglycerides, and lower HDL-cholesterol.
Catalog No. 50-256-2993 Supplier AdipoGen Supplier No. AG40A0112C010
Protein. Human FTO aa 2-505 is fused at the N-terminus to a His-tag. Source E. coli. Endotoxin content <1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in 55mM TRIS-Cl, pH 8.2, containing 150mM NaCl. Purity >90% SDS-PAGE. FTO Fat mass-and obesity-associated gene is the responsible gene for mouse ‘fused toes’ mutation. An association between FTO genotype and type 2 diabetes has been confirmed. The presence of the FTO rs9939609 A-allele was found to be positively correlated with other symptoms of the metabolic syndrome, including higher fasting insulin, glucose, triglycerides, and lower HDL-cholesterol.