Learn More
Sterlitech Corporation FTSH2O Flat Sheet Membrane, CTA, FO, 305 x 305mm, 5/Pk
SDP
Catalog No. 501952328
Encompass_Preferred
Sterlitech offers two brands of forward osmosis flat sheet membranes for separations requiring low energy and low pressure: FTS H2O™ and Aquaporin Inside™*.
FTS H2O™ flat sheet membranes are composed of cellulose and are low-fouling. They are able to process precipitating salts and polymerized organics with minimal impact on the process. 
The Aquaporin Inside™ technology is based on the aquaporin protein--nature's own selective and extremely effective water channel. Aquaporin water channels have been developed through billions of years of evolution, and are now ready to revolutionize industrial water treatment.
Catalog No. 50-195-2328 Supplier Sterlitech Corporation Supplier No. 1121819
Sterlitech offers two brands of forward osmosis flat sheet membranes for separations requiring low energy and low pressure: FTS H2O™ and Aquaporin Inside™*.
FTS H2O™ flat sheet membranes are composed of cellulose and are low-fouling. They are able to process precipitating salts and polymerized organics with minimal impact on the process. 
The Aquaporin Inside™ technology is based on the aquaporin protein--nature's own selective and extremely effective water channel. Aquaporin water channels have been developed through billions of years of evolution, and are now ready to revolutionize industrial water treatment.