Learn More
Aviva Systems Biology Corp FTSJ1 Antibody - N-terminal region (ARP49131_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49131_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FTSJ1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FTSJ1. Alternate names: CDLIV, JM23, MRX44, MRX9, SPB1, TRM7ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-60028