Learn More
Aviva Systems Biology Corp Ftsj3 Antibody - C-terminal region (ARP48805_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48805_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ftsj3. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: Ftsj3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-379