Learn More
Aviva Systems Biology Corp FTSJD1 Antibody - middle region (ARP57215_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57215_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FLJ11171. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FTSJD1. Alternate names: AFT, FTSJD1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53940