Learn More
Medchemexpress LLC FtsZ E.coli His-SU 10ug
SDP
Catalog No. 5000195873
Encompass_Preferred
Recombinant Escherichia coli FtsZ protein, expressed with an N-terminal SUMO and 6xHis affinity tag, supplied lyophilized for biochemical and biophysical studies including polymerization assays and inhibitor screening. The material is characterized by a certificate of analysis and intended for research use only.
  • Recombinant Escherichia coli FtsZ with N-terminal SUMO and 6xHis tag.
  • High purity (>90%) as assessed by reducing SDS-PAGE.
  • Approximate molecular weight 56.3 kDa.
  • Lyophilized formulation in PBS with 6% trehalose, pH 7.4.
  • Recommended reconstitution concentration ≥100 μg/mL in ddH2O.
  • Stable long term at -20 °C; aliquot and store at -80 °C for extended storage.
Catalog No. 50-001-95873 Supplier Medchemexpress LLC Supplier No. HYP7149510UG
Recombinant Escherichia coli FtsZ protein, expressed with an N-terminal SUMO and 6xHis affinity tag, supplied lyophilized for biochemical and biophysical studies including polymerization assays and inhibitor screening. The material is characterized by a certificate of analysis and intended for research use only.
  • Recombinant Escherichia coli FtsZ with N-terminal SUMO and 6xHis tag.
  • High purity (>90%) as assessed by reducing SDS-PAGE.
  • Approximate molecular weight 56.3 kDa.
  • Lyophilized formulation in PBS with 6% trehalose, pH 7.4.
  • Recommended reconstitution concentration ≥100 μg/mL in ddH2O.
  • Stable long term at -20 °C; aliquot and store at -80 °C for extended storage.