Learn More
Medchemexpress LLC FtsZ E.coli His-SU 50ug
SDP
Catalog No. 5000196689
Encompass_Preferred
FtsZ Protein from Escherichia coli, expressed with an N-terminal His-SUMO tag, is a recombinant GTPase used to study bacterial cytoskeletal dynamics and cell division. It is supplied as a purified protein suitable for in vitro biochemical and biophysical assays of polymerization, GTPase activity, and interaction with regulatory factors. Storage and handling recommendations support experimental reproducibility.
Catalog No. 50-001-96689 Supplier Medchemexpress LLC Supplier No. HYP7149550UG
FtsZ Protein from Escherichia coli, expressed with an N-terminal His-SUMO tag, is a recombinant GTPase used to study bacterial cytoskeletal dynamics and cell division. It is supplied as a purified protein suitable for in vitro biochemical and biophysical assays of polymerization, GTPase activity, and interaction with regulatory factors. Storage and handling recommendations support experimental reproducibility.