Learn More
HARVARD BIOSCIENCE INC FTTING AND TUBING KIT
Catalog No. NC0386311
Encompass_Preferred
Total order minimum requirement of $500. If only item on order, must order QTY of 2.Warner Instruments offers a complete tubing and connector kit. This kit is ideal for use with Warner's Imaging and Recording Chambers. The kit is comprised of barbed and Luer fittings, blunt end needles, and an assortment of PE and C-Flex tubing. All barbed and Luer fittings are made from polypropylene and include tube to tube, reducing, Y-, T-, Luer-to-Luer, and Luer-to-barb adapters.
Catalog No. NC0386311 Supplier HARVARD BIOSCIENCE INC Supplier No. 641565
Total order minimum requirement of $500. If only item on order, must order QTY of 2.Warner Instruments offers a complete tubing and connector kit. This kit is ideal for use with Warner's Imaging and Recording Chambers. The kit is comprised of barbed and Luer fittings, blunt end needles, and an assortment of PE and C-Flex tubing. All barbed and Luer fittings are made from polypropylene and include tube to tube, reducing, Y-, T-, Luer-to-Luer, and Luer-to-barb adapters.