Learn More
AVANTOR MASTERFLEX (VWR LLC) FTTNG PVDF S HOSE BARB 10/PK
Catalog No. NC2338474
Encompass
FITTING PVDF STRAIGHT HOSE BARB REDUCER 1/4"ID X1/8"ID 10/PK
Catalog No. NC2338474 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4061456
FITTING PVDF STRAIGHT HOSE BARB REDUCER 1/4"ID X1/8"ID 10/PK