Learn More
Enzo Life Sciences FTY-720. HCl (5 mg). CAS: 162359-56-0
SDP
Supplier:  Enzo Life Sciences BMLSL2330005
Encompass_Preferred
Novel immune modulator that alters migration and homing of lymphocytes. Acts as a potent agonist of sphingosine 1-phosphate receptors (S1P) after phosphorylation by sphingosine kinase. Also inhibits cytosolic phospholipase A2 independently of sphingosine 1-phosphate receptors. Alternative name: 2-Amino-2-[2-(4-octylphenyl)ethyl]-1,3-propanediol . HCl, Fingolimod . HCl. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (10mg/ml), 100% ethanol or dimethyl formamide. Long Term Storage: -20°C.
Catalog No. 50-201-2144