Learn More
Echelon Biosciences Research Labs FTY720 10 mg
SDP
Supplier:  Echelon Biosciences Research Labs B072010MG
Encompass
FTY720 is a structural analog of sphingosine as well as a substrate for sphingosine kinase. The phosphorylated FTY720 acts as a potent agonist at four of the sphingosine-1-phosphate (S1P) receptors (S1P1, S1P3, S1P4, and S1P5). It is a novel immune modulator that prolongs allograft transplant survival in numerous models by inhibiting lymphocyte emigration from lymphoid organs. Animal studies suggest that FTY720 can effectively treat several autoimmune diseases and it has been approved for treating relapsing-remitting multiple sclerosis under the brand name Gilenya (Fingolimod). This item is non returnable
Catalog No. 50-720-021