Learn More
AdipoGen FTY720 . hydrochloride (25 mg)
SDP
Supplier:  AdipoGen AGCR13587M025
Encompass_Preferred
Chemical. CAS: 162359-56-0. Formula: C19H33NO2 . HCl. MW: 307.4 . 36.5. Synthetic analog of ISP-1, isolated from Isaria sinclairii. Cell permeable immunosuppressor displaying lymphocyte sequestration properties and used for treatment in multiple sclerosis. Potent sphingosine 1-phosphate (S1P) receptor (S1P1, S1P3, S1P4, and S1P5) agonist when phosphorylated by sphingosine kinase. Sphingosine transporter Abcb1 and leukotriene C4 transporter Abcc1 activity enhancer. Cytosolic phospholipase A2 inhibitor, independent of sphingosine 1-phosphate receptors. Autophagy inducer. Apoptosis inducer.
Catalog No. 50-205-3954