Learn More
Cayman Chemical FTY720 Phosphate
SDP
Supplier:  Cayman Chemical 1000863910 MG
Encompass_Preferred
A potent agonist at four of the sphingosine-1-phosphate (S1P) receptors with IC50 values of 0.2-6 nM
Catalog No. 50-288-7248