Learn More
Cayman Chemical FTY720 Phosphate
SDP
Catalog No. 502891453
Encompass_Preferred
A potent agonist at four of the sphingosine-1-phosphate (S1P) receptors with IC50 values of 0.2-6 nM
Catalog No. 50-289-1453 Supplier Cayman Chemical Supplier No. 100086391 MG
A potent agonist at four of the sphingosine-1-phosphate (S1P) receptors with IC50 values of 0.2-6 nM