Learn More
Medchemexpress LLC FTY720 (S)-Phosphate | 402616-26-6 | C19H34NO5P | 50 MG
SDP
Catalog No. 5000376036
Encompass_Preferred
FTY720 (S)-Phosphate is an agonist of S1P receptor 1 (S1PR1), used in the research of acute inflammatory diseases such as acute lung injury.
  • Agonist of S1P1 receptor.
  • Maintains S1PR1 protein expression.
  • Enhances human pulmonary artery endothelial cell barrier.
  • Reduces β-arrestin recruitment to S1PR1.
  • Preserves S1PR1 expression in mouse lungs.
  • Protective in bleomycin-induced acute lung injury.
  • Attenuates lung tissue leukocyte infiltration in bleomycin-injured mice.
Catalog No. 50-003-76036 Supplier Medchemexpress LLC Supplier No. HY1538250MG
FTY720 (S)-Phosphate is an agonist of S1P receptor 1 (S1PR1), used in the research of acute inflammatory diseases such as acute lung injury.
  • Agonist of S1P1 receptor.
  • Maintains S1PR1 protein expression.
  • Enhances human pulmonary artery endothelial cell barrier.
  • Reduces β-arrestin recruitment to S1PR1.
  • Preserves S1PR1 expression in mouse lungs.
  • Protective in bleomycin-induced acute lung injury.
  • Attenuates lung tissue leukocyte infiltration in bleomycin-injured mice.