Learn More
Medchemexpress LLC FTY720 (S)-phosphate | 402616-26-6 | MFCD09840646 | 99.6% | 387.45 g/mol | C19H34NO5P | 10 MG
SDP
Catalog No. 5000286097
Encompass_Preferred
FTY720 (S)-Phosphate is the S-stereoisomer of fingolimod phosphorylated at the primary alcohol. It functions as a potent agonist of the sphingosine-1-phosphate receptor 1 (S1PR1) and is used in preclinical research of S1P signaling and acute inflammatory conditions such as acute lung injury.
  • Potent S1PR1 agonist relevant for S1P signaling studies.
  • Single stereoisomer for stereospecific activity.
  • High reported purity suitable for biochemical assays.
  • Stable powder with long-term storage at -20°C.
  • Available in multiple small-scale pack sizes for research use.
Catalog No. 50-002-86097 Supplier Medchemexpress LLC Supplier No. HY1538210MG
FTY720 (S)-Phosphate is the S-stereoisomer of fingolimod phosphorylated at the primary alcohol. It functions as a potent agonist of the sphingosine-1-phosphate receptor 1 (S1PR1) and is used in preclinical research of S1P signaling and acute inflammatory conditions such as acute lung injury.
  • Potent S1PR1 agonist relevant for S1P signaling studies.
  • Single stereoisomer for stereospecific activity.
  • High reported purity suitable for biochemical assays.
  • Stable powder with long-term storage at -20°C.
  • Available in multiple small-scale pack sizes for research use.