Learn More
Aviva Systems Biology Corp FUBP1 Antibody - N-terminal region (ARP37732_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37732_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against FUBP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FUBP1. Alternate names: FBP, FUBPApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56412