Learn More
Aviva Systems Biology Corp FUBP1 Antibody - middle region (ARP35704_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35704_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FUBP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FUBP1. Alternate names: FBP, FUBPApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55823