Learn More
Medchemexpress LLC HY-100758 10mg Medchemexpress, FUBP1–IN-1 CAS:883003-62-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10075810MG
Encompass_Preferred
Medchemexpress, HY-100758 10mg FUBP1–IN-1 CAS:883003-62-1 FUBP1–IN-1 is a potent FUSE binding protein 1 (FUBP1) inhibitor which interferes with the binding of FUBP1 to its single stranded target DNA FUSE sequence , with an IC50 value of 11.0 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7779