Learn More
Medchemexpress LLC FUBP1-IN-1 (FUSE binding protein 1 inhibitor) | 883003-62-1 | 99.7% | 405.39 g·mol⁻¹ | C19H14F3N3O2S | 5 MG
SDP
Catalog No. 5000212000
Encompass_Preferred
FUBP1-IN-1 is a small-molecule inhibitor that disrupts binding of FUSE binding protein 1 (FUBP1) to its single-stranded target DNA and is used in biochemical and cell-based research to probe FUBP1-related pathways. The compound is supplied as a light yellow to yellow solid with specified storage conditions for stability.
  • Potent FUBP1 inhibition with IC50 of 11.0 μM.
  • High chemical purity (99.7%).
  • Molecular weight 405.39 g·mol⁻¹; formula C19H14F3N3O2S.
  • Solid form, light yellow to yellow appearance.
  • Stable under recommended storage conditions for long-term use.
Catalog No. 50-002-12000 Supplier Medchemexpress LLC Supplier No. HY1007585MG
FUBP1-IN-1 is a small-molecule inhibitor that disrupts binding of FUSE binding protein 1 (FUBP1) to its single-stranded target DNA and is used in biochemical and cell-based research to probe FUBP1-related pathways. The compound is supplied as a light yellow to yellow solid with specified storage conditions for stability.
  • Potent FUBP1 inhibition with IC50 of 11.0 μM.
  • High chemical purity (99.7%).
  • Molecular weight 405.39 g·mol⁻¹; formula C19H14F3N3O2S.
  • Solid form, light yellow to yellow appearance.
  • Stable under recommended storage conditions for long-term use.