Learn More
Aviva Systems Biology Corp FUBP3 Antibody - N-terminal region (ARP51746_P050) | 100uL
SDP
Catalog No. 5001460600
Encompass_Preferred
This is a rabbit polyclonal antibody against FUBP3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FUBP3. Alternate names: FBP3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60600 Supplier Aviva Systems Biology Corp Supplier No. ARP51746_P050
This is a rabbit polyclonal antibody against FUBP3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FUBP3. Alternate names: FBP3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat