Learn More
Electron Microscopy Sciences Fuchs Resenthal Counting Chamber (3720)
SDP
Catalog No. 5027812
Encompass_Preferred
Designed for counting leukocytes, eosinophils and cells in cerebrospinal fluid. Grid pattern is etched directly onto the glass surface. The chamber is 0.2 mm deep. Set includes one chamber and 2 octagonal cover glasses. Chrome on Glass.
Catalog No. 50-278-12 Supplier Electron Microscopy Sciences Supplier No. 6351210
Designed for counting leukocytes, eosinophils and cells in cerebrospinal fluid. Grid pattern is etched directly onto the glass surface. The chamber is 0.2 mm deep. Set includes one chamber and 2 octagonal cover glasses. Chrome on Glass.