Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fuchsin Basic (Certified Biological Stain), Fisher Chemical

Catalog No. F9810 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F98-10 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F98-10

missing translation for 'validMainframeMsg'

For use in histology, Feulgen reactions, and bacteriology (staining and in media)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 632-99-5
Molecular Formula C20H20ClN3
Molecular Weight (g/mol) 337.85
MDL Number MFCD00012569
InChI Key AXDJCCTWPBKUKL-UHFFFAOYSA-N
Synonym Magenta, Pararosaniline
PubChem CID 12447
IUPAC Name hydrogen 4-[(4-aminophenyl)(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]aniline chloride
SMILES [H+].[Cl-].CC1=CC(C=CC1=N)=C(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1

Specifications

Melting Point 250°C
Bio. Stain Comm. Certification Pass Test
Dye Content Report %
Grade Certified Biological Stain
Packaging Glass Bottle
Color Index No. 42510
Color Green
pH 5 to 6
Quantity 10 g
Physical Form Solid
Chemical Name or Material Fuchsin Basic
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:2
Flammability:1
Instability:1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.