Learn More
Apexbio Technology LLC Fuchsine base monohydrochloride 632-99-5 10g
SDP
Catalog No. 5000093576
Encompass_Preferred
Fuchsine base monohydrochloride (632-99-5) is a synthetic dye belonging to the triphenylmethane family It is designed for staining applications to visualize cellular and tissue components in histological and cytological studies Fuchsine base monohydrochloride exerts its biological activity primarily through binding with negatively charged groups particularly carboxyl-rich sites within cell structures Based on these properties Fuchsine base monohydrochloride holds research potential in biomedical cytological and histological applications for the analysis of tissue morphology cellular anatomy and biological process visualization
Catalog No. 50-000-93576 Supplier Apexbio Technology LLC Supplier No. A477810000
Fuchsine base monohydrochloride (632-99-5) is a synthetic dye belonging to the triphenylmethane family It is designed for staining applications to visualize cellular and tissue components in histological and cytological studies Fuchsine base monohydrochloride exerts its biological activity primarily through binding with negatively charged groups particularly carboxyl-rich sites within cell structures Based on these properties Fuchsine base monohydrochloride holds research potential in biomedical cytological and histological applications for the analysis of tissue morphology cellular anatomy and biological process visualization