Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fuchsine, Basic, TCI America™
SDP

Catalog No. F009725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F009725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F009725G Supplier TCI America Supplier No. F009725G

missing translation for 'validMainframeMsg'

Chemical Identifiers

Synonym Basic Violet 14

Specifications

UN Number 2811
Color Green
Quantity 25 g
Physical Form Crystal/Powder
Chemical Name or Material Fuchsine, Basic
EINECSNumber (5)-1976
RTECSNumber ZE9705000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.