Learn More
Apexbio Technology LLC Fucoidan 9072-19-9 250mg
SDP
Catalog No. 5000089134
Encompass_Preferred
Fucoidan (CAS 9072-19-9) is a sulfated polysaccharide derived from brown seaweed investigated for its diverse biological activities including anticancer antiviral neuroprotective and immunomodulatory effects Mechanistically fucoidan promotes apoptosis in cancer cells as observed in PC-3 human prostate cancer cells where it activates intrinsic and extrinsic apoptotic pathways via modulation of MAPK (p38 ERK1/2) and PI3K/Akt signaling In vivo studies using murine breast cancer models show that fucoidan suppresses tumor growth inhibits angiogenesis by downregulating VEGF and reduces metastasis These findings support its utility in oncology and immunological research
Catalog No. 50-000-89134 Supplier Apexbio Technology LLC Supplier No. C4038250
Fucoidan (CAS 9072-19-9) is a sulfated polysaccharide derived from brown seaweed investigated for its diverse biological activities including anticancer antiviral neuroprotective and immunomodulatory effects Mechanistically fucoidan promotes apoptosis in cancer cells as observed in PC-3 human prostate cancer cells where it activates intrinsic and extrinsic apoptotic pathways via modulation of MAPK (p38 ERK1/2) and PI3K/Akt signaling In vivo studies using murine breast cancer models show that fucoidan suppresses tumor growth inhibits angiogenesis by downregulating VEGF and reduces metastasis These findings support its utility in oncology and immunological research