Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (-)-Fucose | 2438-80-4 | 164.16 | 1 G
SDP

Catalog No. 5000415645
Encompass_Preferred

(-)-Fucose is classified as a member of the hexoses, playing a role in A and B blood group antigen substructure determination, selectin-mediated leukocyte-endothelial adhesion, and host-microbe interactions. It is orally active and has been shown to inhibit CL11-induced inflammatory responses in the kidney and tumor growth.

  • Endogenous metabolite
  • Orally active
  • Inhibits CL11-induced inflammatory response in kidney
  • Inhibits tumor growth
  • Purity: 98.0%
  • Solid appearance, white to off-white color
  • Classified as a monosaccharide and saccharide
  • Source from human gut microbiota metabolites

Catalog No. 50-004-15645 Supplier Medchemexpress LLC Supplier No. HYN14801G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(-)-Fucose is classified as a member of the hexoses, playing a role in A and B blood group antigen substructure determination, selectin-mediated leukocyte-endothelial adhesion, and host-microbe interactions. It is orally active and has been shown to inhibit CL11-induced inflammatory responses in the kidney and tumor growth.

  • Endogenous metabolite
  • Orally active
  • Inhibits CL11-induced inflammatory response in kidney
  • Inhibits tumor growth
  • Purity: 98.0%
  • Solid appearance, white to off-white color
  • Classified as a monosaccharide and saccharide
  • Source from human gut microbiota metabolites

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.