Learn More
Medchemexpress LLC (-)-Fucose | 2438-80-4 | 164.16 | 10 G
SDP
Catalog No. 5000415638
Encompass_Preferred
(-)-Fucose is a hexose that plays crucial roles in various biological processes, including blood group antigen determination, cell adhesion, and host-microbe interactions. It is known for its oral activity and ability to mitigate inflammatory responses and inhibit tumor growth.
  • Classified as a hexose
  • Plays a role in A and B blood group antigen substructure determination
  • Involved in selectin-mediated leukocyte-endothelial adhesion
  • Contributes to host-microbe interactions
  • Orally active
  • Inhibits CL11-induced inflammatory response in the kidney
  • Inhibits tumor growth
Catalog No. 50-004-15638 Supplier Medchemexpress LLC Supplier No. HYN148010G
(-)-Fucose is a hexose that plays crucial roles in various biological processes, including blood group antigen determination, cell adhesion, and host-microbe interactions. It is known for its oral activity and ability to mitigate inflammatory responses and inhibit tumor growth.
  • Classified as a hexose
  • Plays a role in A and B blood group antigen substructure determination
  • Involved in selectin-mediated leukocyte-endothelial adhesion
  • Contributes to host-microbe interactions
  • Orally active
  • Inhibits CL11-induced inflammatory response in the kidney
  • Inhibits tumor growth