Learn More
Medchemexpress LLC (-)-Fucose | 2438-80-4 | 164.16 | 5 G
SDP
Catalog No. 5000416391
Encompass_Preferred
(-)-Fucose is classified as a hexose and plays a role in the determination of A and B blood group antigen substructures, selectin-mediated leukocyte-endothelial adhesion, and host-microbe interactions. It is orally active and inhibits CL11-induced inflammatory responses in the kidney and tumor growth.
  • Endogenous metabolite
  • Plays a role in A and B blood group antigen substructure determination
  • Involved in selectin-mediated leukocyte-endothelial adhesion
  • Contributes to host-microbe interactions
  • Orally active
  • Inhibits CL11-induced inflammatory response in kidney
  • Inhibits tumor growth
Catalog No. 50-004-16391 Supplier Medchemexpress LLC Supplier No. HYN14805G
(-)-Fucose is classified as a hexose and plays a role in the determination of A and B blood group antigen substructures, selectin-mediated leukocyte-endothelial adhesion, and host-microbe interactions. It is orally active and inhibits CL11-induced inflammatory responses in the kidney and tumor growth.
  • Endogenous metabolite
  • Plays a role in A and B blood group antigen substructure determination
  • Involved in selectin-mediated leukocyte-endothelial adhesion
  • Contributes to host-microbe interactions
  • Orally active
  • Inhibits CL11-induced inflammatory response in kidney
  • Inhibits tumor growth