Learn More
Medchemexpress LLC (-)-Fucose | 2438-80-4 | 164.16 | 500 MG
SDP
Catalog No. 5000414871
Encompass_Preferred
(-)-Fucose is a hexose involved in A and B blood group antigen substructure determination, selectin-mediated leukocyte-endothelial adhesion, and host-microbe interactions. It is orally active and inhibits CL11-induced inflammatory response in the kidney and tumor growth.
  • Classified as a hexose.
  • Plays a role in A and B blood group antigen substructure determination.
  • Involved in selectin-mediated leukocyte-endothelial adhesion.
  • Participates in host-microbe interactions.
  • Orally active.
  • Inhibits CL11-induced inflammatory response in the kidney.
  • Inhibits tumor growth.
  • Purity of 98.0%.
  • White to off-white solid appearance.
Catalog No. 50-004-14871 Supplier Medchemexpress LLC Supplier No. HYN1480500MG
(-)-Fucose is a hexose involved in A and B blood group antigen substructure determination, selectin-mediated leukocyte-endothelial adhesion, and host-microbe interactions. It is orally active and inhibits CL11-induced inflammatory response in the kidney and tumor growth.
  • Classified as a hexose.
  • Plays a role in A and B blood group antigen substructure determination.
  • Involved in selectin-mediated leukocyte-endothelial adhesion.
  • Participates in host-microbe interactions.
  • Orally active.
  • Inhibits CL11-induced inflammatory response in the kidney.
  • Inhibits tumor growth.
  • Purity of 98.0%.
  • White to off-white solid appearance.