Learn More
Medchemexpress LLC (-)-Fucose (6-Desoxygalactose) | 2438-80-4 | 164.16 | 1 ML
SDP
Catalog No. 5000416390
Encompass_Preferred
(-)-Fucose is a hexose that plays a role in various biological processes. It is involved in A and B blood group antigen substructure determination, selectin-mediated leukocyte-endothelial adhesion, and host-microbe interactions. This compound is orally active and has been shown to inhibit CL11-induced inflammatory responses in the kidney and suppress tumor growth. This product is intended for research use only.
  • Plays a role in A and B blood group antigen substructure determination
  • Involved in selectin-mediated leukocyte-endothelial adhesion
  • Participates in host-microbe interactions
  • Is orally active
  • Inhibits CL11-induced inflammatory response in kidney
  • Inhibits tumor growth
Catalog No. 50-004-16390 Supplier Medchemexpress LLC Supplier No. HYN148010MM/1ML
(-)-Fucose is a hexose that plays a role in various biological processes. It is involved in A and B blood group antigen substructure determination, selectin-mediated leukocyte-endothelial adhesion, and host-microbe interactions. This compound is orally active and has been shown to inhibit CL11-induced inflammatory responses in the kidney and suppress tumor growth. This product is intended for research use only.
  • Plays a role in A and B blood group antigen substructure determination
  • Involved in selectin-mediated leukocyte-endothelial adhesion
  • Participates in host-microbe interactions
  • Is orally active
  • Inhibits CL11-induced inflammatory response in kidney
  • Inhibits tumor growth