Learn More
Bioworld FUCOSE SEPAROPOREAGAROSE)
Supplier:  Bioworld 201810101
Encompass
FUCOSE SEPAROPORE (AGAROSE) 4B-CL: Affinity Matrix for fucose-binding glycoproteins. Fucose is linked to Separopore4B-CL (fastflow)via a spacer arm to ensure effficient binding of this ligand to the protein. Specifications: Molecular W...
Catalog No. 50-197-683