Learn More
Medchemexpress LLC (-)-Fucose (Standard) | 2438-80-4 | 164.16 | 25 MG
SDP
Catalog No. 5000390318
Encompass_Preferred
(-)-Fucose (Standard) is the analytical standard of (-)-Fucose, intended for research and analytical applications. It is classified as a hexose and plays a role in A and B blood group antigen substructure determination, selectin-mediated leukocyte-endothelial adhesion, and host-microbe interactions. (-)-Fucose is orally active and has been shown to inhibit CL11-induced inflammatory response in the kidney and tumor growth.
  • Analytical standard for (-)-Fucose
  • Intended for research and analytical applications
  • Plays a role in A and B blood group antigen substructure determination
  • Involved in selectin-mediated leukocyte-endothelial adhesion
  • Participates in host-microbe interactions
  • Orally active
  • Inhibits CL11-induced inflammatory response in kidney and tumor growth
  • Purity: 99.88%
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-003-90318 Supplier Medchemexpress LLC Supplier No. 25MGHYN1480R
(-)-Fucose (Standard) is the analytical standard of (-)-Fucose, intended for research and analytical applications. It is classified as a hexose and plays a role in A and B blood group antigen substructure determination, selectin-mediated leukocyte-endothelial adhesion, and host-microbe interactions. (-)-Fucose is orally active and has been shown to inhibit CL11-induced inflammatory response in the kidney and tumor growth.
  • Analytical standard for (-)-Fucose
  • Intended for research and analytical applications
  • Plays a role in A and B blood group antigen substructure determination
  • Involved in selectin-mediated leukocyte-endothelial adhesion
  • Participates in host-microbe interactions
  • Orally active
  • Inhibits CL11-induced inflammatory response in kidney and tumor growth
  • Purity: 99.88%
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS