Learn More
TARGETMOL CHEMICALS INC FUCOSTEROL 10MG
SDP
Catalog No. 502579097
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Fucosterol is isolated from E. stolonifera with anti-diabetic anti-adipogenic and anti-cancer activities. It regulates adipogenesis via modulation of PPAR(alpha) and C/EBP(alpha) expression. purity: 99%
Catalog No. 50-257-9097 Supplier TARGETMOL CHEMICALS INC Supplier No. T818410MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Fucosterol is isolated from E. stolonifera with anti-diabetic anti-adipogenic and anti-cancer activities. It regulates adipogenesis via modulation of PPAR(alpha) and C/EBP(alpha) expression. purity: 99%