Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbosynth LLC. Fucosyl-GM1 Ganglioside, 2 mg, White Off-White Powder, Min. 98 Area-% Purity
SDP

Catalog No. NC3252590
Encompass_Preferred

Fucosyl-GM1 Ganglioside is a notable monosialylated glycosphingolipid sourced from BIOSYNTH INTERNATIONAL INC. It showcases minimal expression in healthy tissues but has a significant prevalence in certain tumors, such as small cell lung cancer (SCLC), positioning it as an important potential tumor marker and antigen target for immunotherapy. This product is strictly intended for research purposes only.

  • CAS Number: 71812-11-8
  • Synonyms: Fucosylated monosialoganglioside GM1, Fuc-GM1, Fucosyl GM1, Ganglioside FG1
  • Molecular Weight: 1692 g/mol
  • Molecular Formula: C79H141N3O35
  • Purity: Min. 98 Area-%
  • Appearance: White off-white powder
  • Storage Conditions: Store at less than -15 °C, close container well
  • Research Use: For research purposes only
  • Biological Role: Potential tumor marker for SCLC
  • Therapeutic Potential: Potential antigen target for immunotherapy

Catalog No. NC3252590 Supplier Carbosynth LLC. Supplier No. OF633662MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fucosyl-GM1 Ganglioside is a notable monosialylated glycosphingolipid sourced from BIOSYNTH INTERNATIONAL INC. It showcases minimal expression in healthy tissues but has a significant prevalence in certain tumors, such as small cell lung cancer (SCLC), positioning it as an important potential tumor marker and antigen target for immunotherapy. This product is strictly intended for research purposes only.

  • CAS Number: 71812-11-8
  • Synonyms: Fucosylated monosialoganglioside GM1, Fuc-GM1, Fucosyl GM1, Ganglioside FG1
  • Molecular Weight: 1692 g/mol
  • Molecular Formula: C79H141N3O35
  • Purity: Min. 98 Area-%
  • Appearance: White off-white powder
  • Storage Conditions: Store at less than -15 °C, close container well
  • Research Use: For research purposes only
  • Biological Role: Potential tumor marker for SCLC
  • Therapeutic Potential: Potential antigen target for immunotherapy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.