Learn More
Cayman Chemical Fucoxnthn 10mg
SDP
Catalog No. 502953209
Encompass_Preferred
A carotenoid that occurs naturally in certain algae; significantly reduces abdominal white adipose tissue (WAT) in mice and rats when included in their diet; increases the amount of UCP1 protein in WAT; decreases blood glucose and plasma insulin levels in diabetic mice
Catalog No. 50-295-3209 Supplier Cayman Chemical Supplier No. 1306810 MG
A carotenoid that occurs naturally in certain algae; significantly reduces abdominal white adipose tissue (WAT) in mice and rats when included in their diet; increases the amount of UCP1 protein in WAT; decreases blood glucose and plasma insulin levels in diabetic mice