Learn More
Cayman Chemical Fucoxanthin, Purity: ≥95%, CAS: 3351-86-8, 50 mg
SDP
Catalog No. 502955710
Encompass_Preferred
A carotenoid that occurs naturally in certain algae; significantly reduces abdominal white adipose tissue (WAT) in mice and rats when included in their diet; increases the amount of UCP1 protein in WAT; decreases blood glucose and plasma insulin levels in diabetic mice
Catalog No. 50-295-5710 Supplier Cayman Chemical Supplier No. 1306850 MG
A carotenoid that occurs naturally in certain algae; significantly reduces abdominal white adipose tissue (WAT) in mice and rats when included in their diet; increases the amount of UCP1 protein in WAT; decreases blood glucose and plasma insulin levels in diabetic mice