Learn More
Medchemexpress LLC HY-B0393 100mg Medchemexpress, Fudosteine CAS:13189-98-5 Purity:>98%
SDP
Catalog No. 502082515
Encompass_Preferred
Medchemexpress, HY-B0393 100mg Fudosteine CAS:13189-98-5 Fudosteine is a cysteine derivative and a mucoactive agent. Fudosteine inhibits MUC5AC mucin hypersecretion by reducing MUC5AC gene expression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2515 Supplier Medchemexpress LLC Supplier No. HYB0393100MG
Medchemexpress, HY-B0393 100mg Fudosteine CAS:13189-98-5 Fudosteine is a cysteine derivative and a mucoactive agent. Fudosteine inhibits MUC5AC mucin hypersecretion by reducing MUC5AC gene expression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.