Learn More
Bioworld FUDR, 1 g
Catalog No. 50256011
Encompass_Preferred
Inhibits DNA synthesis by prior inhibition of thymidylate synthetase.An experimental antiproliferative agent shown to have activity against variety of neoplasms Usually ships within 24 hours. Ths product is sold for laboratory research use only.RTECS Number: YU7525000
Catalog No. 50-256-011 Supplier Bioworld Supplier No. 406900161
Inhibits DNA synthesis by prior inhibition of thymidylate synthetase.An experimental antiproliferative agent shown to have activity against variety of neoplasms Usually ships within 24 hours. Ths product is sold for laboratory research use only.RTECS Number: YU7525000