Learn More
Medchemexpress LLC HY-109059 10mg Medchemexpress, Fulacimstat CAS:1488354-15-9 Purity:>98%
SDP
Catalog No. 501968663
Encompass_Preferred
Medchemexpress, HY-109059 10mg Fulacimstat CAS:1488354-15-9 Fulacimstat is an orally available chymase inhibitor, with IC50s of 4, 3 nM for human and hamster chymase enzyme, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8663 Supplier Medchemexpress LLC Supplier No. HY10905910MG
Medchemexpress, HY-109059 10mg Fulacimstat CAS:1488354-15-9 Fulacimstat is an orally available chymase inhibitor, with IC50s of 4, 3 nM for human and hamster chymase enzyme, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.